accessibility menu, dialog, popup

UserName

H3COCH3O(Z)

CAS RN 3681-71-8

IUPAC Name: (3Z)-hex-3-en-1-yl acetate
Synonyms: (Z)-3-hexenyl acetate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: NPFVOOAXDOBMCE-PLNGDYQASA-N
SMILES: CC\C=C/CCOC(C)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results