missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexenyl acetate, 98+%

Catalog No. AAA1938022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1938022 100 g
AAA1938014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1938022 Supplier Thermo Scientific Chemicals Supplier No. A1938022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3681-71-8 | C8H14O2 | 142.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3681-71-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00036563
InChI Key NPFVOOAXDOBMCE-PLNGDYQASA-N
Synonym cis-3-hexenyl acetate, z-3-hexenyl acetate, z-hex-3-enyl acetate, 3-hexenylacetate, cis-3-hexenyl ethanoate, cis-3-hexen-1-yl acetate, leaf acetate, 3-hexen-1-ol, acetate, z, cis-3-hexenol acetate, 3z-hex-3-en-1-yl acetate
PubChem CID 5363388
ChEBI CHEBI:61316
IUPAC Name [(Z)-hex-3-enyl] acetate
SMILES CCC=CCCOC(=O)C

Specifications

Density 0.899
Boiling Point 169°C
Flash Point 59°C (138°F)
Refractive Index 1.427
Quantity 100 g
UN Number UN3272
Beilstein 1721854
Formula Weight 142.2
Percent Purity 99%
Chemical Name or Material cis-3-Hexenyl acetate
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 222-960-1
RTECSNumber MP8425050
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only