missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexenyl Acetate 97.0+%, TCI America™

Catalog No. A088825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A088825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A088825ML Supplier TCI America Supplier No. A088825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3681-71-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00036563
InChI Key NPFVOOAXDOBMCE-PLNGDYQASA-N
Synonym cis-3-hexenyl acetate, z-3-hexenyl acetate, z-hex-3-enyl acetate, 3-hexenylacetate, cis-3-hexenyl ethanoate, cis-3-hexen-1-yl acetate, leaf acetate, 3-hexen-1-ol, acetate, z, cis-3-hexenol acetate, 3z-hex-3-en-1-yl acetate
PubChem CID 5363388
ChEBI CHEBI:61316
IUPAC Name [(Z)-hex-3-enyl] acetate
SMILES CCC=CCCOC(=O)C

Specifications

Color Colorless
Boiling Point 169°C
Quantity 25 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexenyl Acetate
EINECSNumber (2)-0760&(2)-2533
RTECSNumber MP8425050
TSCA Yes