missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,3-Dibromopropionate 98.0+%, TCI America™

Catalog No. D1992500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1992500G 500 g
D199225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1992500G Supplier TCI America Supplier No. D1992500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3674-13-3
Molecular Formula C5H8Br2O2
Molecular Weight (g/mol) 259.925
MDL Number MFCD00000212
InChI Key OENICUBCLXKLJQ-UHFFFAOYSA-N
Synonym ethyl 2,3-dibromopropionate, propanoic acid, 2,3-dibromo-, ethyl ester, ethyl alpha,beta-dibromopropionate, propionic acid, 2,3-dibromo-, ethyl ester, ethyl-2,3-dibromopropionate, 2,3-dibromopropionic acid ethyl ester, ethyl .alpha.,.beta.-dibromopropionate, propionic acid, 2,3-dibromo-, ethyl ester 6ci,7ci,8ci, ethyl2,3-dibromopropionate, acmc-1ahn6
PubChem CID 97945
IUPAC Name ethyl 2,3-dibromopropanoate
SMILES CCOC(=O)C(CBr)Br

Specifications

Color Yellow
Boiling Point 82°C
Quantity 500 g
UN Number 2922
Formula Weight 259.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2,3-Dibromopropionate
RTECSNumber UA2458382
TSCA Yes