accessibility menu, dialog, popup

UserName

CAS RN 367-24-8

IUPAC Name: 4-bromo-2-fluoroaniline
Synonyms: 4-bromo-2-fluoroaniline
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: GZRMNMGWNKSANY-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C=C1F
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)