accessibility menu, dialog, popup

UserName

CAS RN 3658-80-8

IUPAC Name: dimethyltrisulfane
Synonyms: dimethyl trisulfide methyl trisulfide (8CI) dimethyl trisulphide methyl trisulfide trisulfide, dimethyl
Molecular Weight (g/mol): 126.25
Molecular Formula: C2H6S3
InChi Key: YWHLKYXPLRWGSE-UHFFFAOYSA-N
SMILES: CSSSC
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)