accessibility menu, dialog, popup

UserName

ONOFF

CAS RN 364-74-9

IUPAC Name: 1,4-difluoro-2-nitrobenzene
Synonyms: benzene, 1,4-difluoro-2-nitro-
Molecular Weight (g/mol): 159.09
Molecular Formula: C6H3F2NO2
InChi Key: XNJAYQHWXYJBBD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results