accessibility menu, dialog, popup

UserName

HO

CAS RN 3637-61-4

IUPAC Name: cyclopentylmethanol
Synonyms: cyclopentanemethanol
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: ISQVBYGGNVVVHB-UHFFFAOYSA-N
SMILES: OCC1CCCC1
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)