missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanemethanol, 98%

Catalog No. AAA1159522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1159522 100 g
AAA1159506 5 g
AAA1159514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1159522 Supplier Thermo Scientific Chemicals Supplier No. A1159522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3637-61-4 | C6H12O | 100.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3637-61-4
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00001384
InChI Key ISQVBYGGNVVVHB-UHFFFAOYSA-N
Synonym cyclopentanemethanol, cyclopentyl-methanol, cyclopentylcarbinol, cyclopentylmethyl alcohol, cyclopentanmethanol, cylcopentanemethanol, cyclopentan-methanol, cyclopentyl methanol, cyclopentyl carbinol, cyclo-pentanemethanol
PubChem CID 77195
IUPAC Name cyclopentylmethanol
SMILES C1CCC(C1)CO

Specifications

Density 0.925
Boiling Point 162°C to 163°C
Flash Point 62°C (143°F)
Refractive Index 1.458
Quantity 100 g
Beilstein 1919000
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material Cyclopentanemethanol
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 222-861-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only