missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanemethanol 98.0+%, TCI America™

Catalog No. C26585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26585G 5 g
C265825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26585G Supplier TCI America Supplier No. C26585G

Chemical Identifiers

CAS 3637-61-4
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00001384
InChI Key ISQVBYGGNVVVHB-UHFFFAOYSA-N
Synonym cyclopentanemethanol, cyclopentyl-methanol, cyclopentylcarbinol, cyclopentylmethyl alcohol, cyclopentanmethanol, cylcopentanemethanol, cyclopentan-methanol, cyclopentyl methanol, cyclopentyl carbinol, cyclo-pentanemethanol
PubChem CID 77195
IUPAC Name cyclopentylmethanol
SMILES C1CCC(C1)CO

Specifications

Color Colorless
Boiling Point 163°C
Quantity 5 g
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentanemethanol