accessibility menu, dialog, popup

UserName

H3COCl

CAS RN 36215-07-3

IUPAC Name: 1-chloro-3-methoxypropane
Synonyms: 1-chloro-3-methoxy-propane
Molecular Weight (g/mol): 108.57
Molecular Formula: C4H9ClO
InChi Key: BQLHMMQUVJCTAN-UHFFFAOYSA-N
SMILES: COCCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results