missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-methoxypropane 98.0+%, TCI America™

Catalog No. C205525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C205525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C205525G Supplier TCI America Supplier No. C205525G
Only null left

Chemical Identifiers

CAS 36215-07-3
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.57
MDL Number MFCD05663419
InChI Key BQLHMMQUVJCTAN-UHFFFAOYSA-N
Synonym 3-Chloropropyl Methyl Ether
PubChem CID 118927
IUPAC Name 1-chloro-3-methoxypropane
SMILES COCCCCl

Specifications

Color Yellow
Boiling Point 114°C
Quantity 25 g
UN Number 1993
Formula Weight 108.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-3-methoxypropane