missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-2,6-dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Chemical Name or Material | 4-Methoxy-2,6-dimethylphenylboronic Acid (contains varying amounts of Anhydride) |
| Melting Point | 175°C |
| CAS | 361543-99-9 |
| Color | White |
| Quantity | 1 g |
| Physical Form | Crystal-Powder at 20°C |
| Molecular Formula | C9H13BO3 |
| MDL Number | MFCD03452755 |
| Synonym | 2,6-dimethyl-4-methoxyphenyl boronic acid, 2,6-dimethyl-4-methoxyphenylboronic acid, 4-methoxy-2,6-dimethylphenyl boronic acid, 2,6-dimethyl-4-methoxybenzeneboronic acid, 2-borono-5-methoxy-m-xylene, 4-borono-3,5-dimethylanisole, 4-methoxy-2,6-dimethylbenzeneboronic acid, 4-methoxy-2,6-dimethylphenylboronicacid, boronic acid, 4-methoxy-2,6-dimethylphenyl, acmc-209il9 |
| InChI Key | UOSSBXMFWPYHEF-UHFFFAOYSA-N |
| Show More |