accessibility menu, dialog, popup

UserName

CAS RN 361346-80-7

CH3CH3CH3CH3H3CNHOCH3OCH3HNOCH3OH3CH3CCo

CAS RN 361346-80-7

IUPAC Name: 3-[({2-[(2-acetyl-3-oxobut-1-en-1-yl)amino]-1,2-bis(2,4,6-trimethylphenyl)ethyl}amino)methylidene]pentane-2,4-dione cobalt
Synonyms: 3-[({2-[(2-acetyl-3-oxobut-1-en-1-yl)amino]-1,2-bis(2,4,6-trimethylphenyl)ethyl}amino)methylidene]pentane-2,4-dione cobalt
Molecular Weight (g/mol): 575.62
Molecular Formula: C32H40CoN2O4
InChi Key: WDSKJOXWWBKMGO-UHFFFAOYNA-N
SMILES: [Co].CC(=O)C(=CNC(C(NC=C(C(C)=O)C(C)=O)C1=C(C)C=C(C)C=C1C)C1=C(C)C=C(C)C=C1C)C(C)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results