missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato Cobalt(II), TCI America™

Catalog No. B2314100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2314100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B2314100MG Supplier TCI America Supplier No. B2314100MG
Only null left

Chemical Identifiers

CAS 361346-80-7
Molecular Formula C32H40CoN2O4
Molecular Weight (g/mol) 575.615
MDL Number MFCD06797061
InChI Key WDSKJOXWWBKMGO-JOVGIXRXSA-N
Synonym (R)-AMAC
PubChem CID 11226930
IUPAC Name 3-[[[(1R,2R)-2-[(2-acetyl-3-oxobut-1-enyl)amino]-1,2-bis(2,4,6-trimethylphenyl)ethyl]amino]methylidene]pentane-2,4-dione;cobalt
SMILES CC1=CC(=C(C(=C1)C)C(C(C2=C(C=C(C=C2C)C)C)NC=C(C(=O)C)C(=O)C)NC=C(C(=O)C)C(=O)C)C.[Co]

Specifications

Quantity 100 mg
Formula Weight 573.60
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-N,N′-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato Cobalt(II)