accessibility menu, dialog, popup

UserName

CH3OH(Z)

CAS RN 35854-86-5

IUPAC Name: (6Z)-non-6-en-1-ol
Synonyms: cis-6-nonenol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: XJHRZBIBSSVCEL-ARJAWSKDSA-N
SMILES: CC\C=C/CCCCCO
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)