missing translation for 'onlineSavingsMsg'
Learn More

cis-6-Nonen-1-ol, 95%

Catalog No. AAL0838706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0838706 5 g
AAL0838703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0838706 Supplier Thermo Scientific Chemicals Supplier No. L0838706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35854-86-5 | C9H18O | 142.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35854-86-5
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00015388
InChI Key XJHRZBIBSSVCEL-ARJAWSKDSA-N
Synonym cis-6-nonen-1-ol, cis-6-nonenol, z-non-6-en-1-ol, z-6-nonen-1-ol, 6z-nonen-1-ol, 6z-non-6-en-1-ol, 6-nonen-1-ol, z, z-6-nonenol, 6z-6-nonen-1-ol
PubChem CID 5362792
IUPAC Name (Z)-non-6-en-1-ol
SMILES CCC=CCCCCCO

Specifications

Density 0.85
Boiling Point 85°C (5 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.451
Quantity 5 g
Beilstein 2322878
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material cis-6-Nonen-1-ol
EINECSNumber 252-764-1
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only