accessibility menu, dialog, popup

UserName

CAS RN 35607-66-0

CH3O(S)HNOS(R)SNOHOOOH2NO

CAS RN 35607-66-0

IUPAC Name: (6R,7S)-3-[(carbamoyloxy)methyl]-7-methoxy-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: cefoxitin mefoxitin
Molecular Weight (g/mol): 427.45
Molecular Formula: C16H17N3O7S2
InChi Key: WZOZEZRFJCJXNZ-ZBFHGGJFSA-N
SMILES: CO[C@]1(NC(=O)CC2=CC=CS2)[C@H]2SCC(COC(N)=O)=C(N2C1=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results