missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cefoxitin, 98%

Catalog No. AAJ6689103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6689103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6689103 Supplier Thermo Scientific Chemicals Supplier No. J6689103
Only null left

CAS: 35607-66-0 | C16H17N3O7S2 | 427.446 g/mol

Cefoxitin is a semisynthetic, broad-spectrum second-generation cephalosporin with antibacterial activity. The activity of cefoxitin results in the weakening of the bacterial cell wall and causes cell lysis. Cefoxitin acts by interfering with cell wall synthesis. Its activity spectrum includes a broad range of gram-negative and gram-positive bacteria including anaerobes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cefoxitin is a semisynthetic, broad-spectrum second-generation cephalosporin with antibacterial activity. The activity of cefoxitin results in the weakening of the bacterial cell wall and causes cell lysis. Cefoxitin acts by interfe

Chemical Identifiers

CAS 35607-66-0
Molecular Formula C16H17N3O7S2
Molecular Weight (g/mol) 427.446
MDL Number MFCD00072014
InChI Key WZOZEZRFJCJXNZ-ZBFHGGJFSA-N
PubChem CID 441199
ChEBI CHEBI:209807
IUPAC Name (6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)O)NC(=O)CC3=CC=CS3

Specifications

Type Cefoxitin
Physical Form Powder
Melting Point 149°C to 150°C
Color White
Quantity 1 g
Flash Point >110°C (230°F)
Boiling Point 843°C
Solubility Information Predicted solubility in water is less than 0.2mg/mL
Formula Weight 427.45
Percent Purity 98%
Chemical Name or Material Cefoxitin
Show More Show Less
Hazard Category H334
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P285-P304+P341-P342+P311-P501c
EINECSNumber 252-641-2
RTECSNumber XI0386500
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only