accessibility menu, dialog, popup

UserName

CAS RN 35543-24-9

ClH3COOCH3ONHOH3C

CAS RN 35543-24-9

IUPAC Name: 1-[4-oxo-4-(2,4,6-trimethoxyphenyl)butyl]pyrrolidin-1-ium chloride
Synonyms: buflomedil
Molecular Weight (g/mol): 343.85
Molecular Formula: C17H26ClNO4
InChi Key: ZDPACSAHMZADFZ-UHFFFAOYSA-N
SMILES: [Cl-].COC1=CC(OC)=C(C(=O)CCC[NH+]2CCCC2)C(OC)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results