missing translation for 'onlineSavingsMsg'
Learn More

Buflomedil hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111014010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014010 Supplier Merck Emd Millipore Supplier No. B58991G

Buflomedil hydrochloride is a vasoactive agent. Its mode of action involves increasing the peripheral and cerebral blood flow in ischaemic tissues of patients, who are mostly affected with vascular diseases.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 35543-24-9
Physical Form Neat
Linear Formula C17H25 NO4 · HCl
MDL Number MFCD00078965
Synonym 4-(1-Pyrrolidinyl)-1-(2,4,6-trimethoxyphenyl)-1-butanone
Type Analytical Standard
Quantity 1 g
Formula Weight 343.85
Grade Analytical Standard