accessibility menu, dialog, popup

UserName

CAS RN 355-93-1

IUPAC Name: 2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate
Synonyms: octafluoropentyl methacrylate
Molecular Weight (g/mol): 300.15
Molecular Formula: C9H8F8O2
InChi Key: ZNJXRXXJPIFFAO-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)