missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate, 98%, stab.

Catalog No. AAL1204414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1204414 25 g
AAL1204406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1204414 Supplier Thermo Scientific Chemicals Supplier No. L1204414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 355-93-1 | C9H8F8O2 | 300.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 355-93-1
Molecular Formula C9H8F8O2
Molecular Weight (g/mol) 300.15
MDL Number MFCD00039278
InChI Key ZNJXRXXJPIFFAO-UHFFFAOYSA-N
Synonym 1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate
PubChem CID 67739
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)F

Specifications

Density 1.432
Boiling Point 178°C to 179°C
Flash Point 81°C (177°F)
Refractive Index 1.36
Quantity 25 g
Beilstein 1801409
Formula Weight 300.15
Percent Purity 98%
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate, stabilized
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 206-596-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only