missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,5H-Octafluoropentyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. O04815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04815G 5 g
O048125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04815G Supplier TCI America Supplier No. O04815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 355-93-1
Molecular Formula C9H8F8O2
Molecular Weight (g/mol) 300.15
MDL Number MFCD00039278
InChI Key ZNJXRXXJPIFFAO-UHFFFAOYSA-N
Synonym 1h,1h,5h-octafluoropentyl methacrylate, 2,2,3,3,4,4,5,5-octafluoropentyl methacrylate, octafluoropentyl methacrylate, 1h,1h,5h-perfluoropentyl methacrylate, 1h,1h,5h-octafluoropentyl methacrylate stabilized with mehq, 2-propenoic acid, 2-methyl-, 2,2,3,3,4,4,5,5-octafluoropentyl ester, methacrylic acid 1h,1h,5h-perfluoropentyl ester, methacrylic acid 1h,1h,5h-octafluoropentyl ester, octafluoropentyl methacrylate polymer, 1h,1h,5h-octafluoropentylmethacrylate
PubChem CID 67739
IUPAC Name 2,2,3,3,4,4,5,5-octafluoropentyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)F

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 g
Formula Weight 300.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,5H-Octafluoropentyl Methacrylate (stabilized with MEHQ)