accessibility menu, dialog, popup

UserName

HClSnClHH3CCH3

CAS RN 3542-36-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 418.07
Molecular Formula: C16H36Cl2Sn
InChi Key: DXPJPPSKHFECLR-UHFFFAOYSA-N
SMILES: Cl.Cl.CCCCCCCC[SnH2]CCCCCCCC
Boiling Point 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)