missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Di-n-octyldichlorotin, 95%

Catalog No. AA4173803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4173803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4173803 Supplier Thermo Scientific™ Supplier No. 4173803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 3542-36-7
Melting Point 44-48°
Boiling Point 175°/1 mm
Molecular Formula [CH{3}(CH{2}){7}]{2}SnCl{2}
MDL Number MFCD00018091
Quantity 1 g
UN Number UN3261
Synonym Dichlorodi-n-octyltin;Dichlorodi-n-octylstannane
Formula Weight 416.04
Percent Purity 95%
Physical Form Solid
Show More Show Less