missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyldichlorotin, 95%

Catalog No. AA4173814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4173814 25 g
AA4173806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4173814 Supplier Thermo Scientific Chemicals Supplier No. 04173814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3542-36-7 | C16H36Cl2Sn | 418.07 g/mol

Di-n-octyldichlorotin compounds are used as intermediates in the production of other organotin chemicals. Metatin and Acima. These products are also used as catalysts or industrial intermediates in the production of polyurethanes, silicone resins, synthetic lubricants, and MS polymer sealants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3542-36-7
Molecular Formula C16H36Cl2Sn
Molecular Weight (g/mol) 418.07
MDL Number MFCD00018091
InChI Key DXPJPPSKHFECLR-UHFFFAOYSA-N
Synonym dichlorodioctyltin, dioctyltin dichloride, di-n-octyltin dichloride, stannane, dichlorodioctyl, dotc, di-n-octyltindichloride, dichloro dioctyl stannane, di-n-octyldichlorotin, stannane, dioctyldichloro, di-n-octyl-zinn dichlorid
PubChem CID 77080
IUPAC Name dichloro(dioctyl)stannane
SMILES Cl.Cl.CCCCCCCC[SnH2]CCCCCCCC

Specifications

Melting Point 44°C to 48°C
Boiling Point 175°C (1 mmHg)
Quantity 25 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 416.04
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Di-n-octyldichlorotin
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 222-583-2
RTECSNumber WH7247000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only