accessibility menu, dialog, popup

UserName

H3COOCH2HNCH3O

CAS RN 35356-70-8

IUPAC Name: methyl 2-acetamidoprop-2-enoate
Synonyms: methyl 2-acetamidoprop-2-enoate
Molecular Weight (g/mol): 143.14
Molecular Formula: C6H9NO3
InChi Key: SMWNFFKPVLVOQQ-UHFFFAOYSA-N
SMILES: COC(=O)C(=C)NC(C)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)