missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Acetamidoacrylate 97.0+%, TCI America™

Catalog No. A13045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13045G 5 g
A13041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13045G Supplier TCI America Supplier No. A13045G

Chemical Identifiers

CAS 35356-70-8
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.14
MDL Number MFCD00013394
InChI Key SMWNFFKPVLVOQQ-UHFFFAOYSA-N
Synonym methyl 2-acetamidoacrylate, 2-acetamidoacrylic acid methyl ester, n-acetyldehydroalanine methyl ester, 2-propenoic acid, 2-acetylamino-, methyl ester, methyl 2-acetylamino acrylate, 2-acetamidomethacrylate, acmc-209igf, methyl 2-acetaminoacrylate, methyl 2-acetylamino-acrylate, methyl 2-acetamido-2-propenate
PubChem CID 98644
IUPAC Name methyl 2-acetamidoprop-2-enoate
SMILES COC(=O)C(=C)NC(C)=O

Specifications

Melting Point 51°C
Color White-Yellow
Boiling Point 104°C
Quantity 5 g
Formula Weight 143.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Acetamidoacrylate
TSCA No
Recommended Storage Refrigerator