missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-acetamidoacrylate, 97%

Catalog No. AAL1004906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1004906 5 g
AAL1004903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1004906 Supplier Thermo Scientific Chemicals Supplier No. L1004906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 35356-70-8 | C6H9NO3 | 143.14 g/mol

Methyl 2-acetamidoacrylate exhibits anti-inflammatory property and decreases induced acute kidney injury in mice.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-acetamidoacrylate exhibits anti-inflammatory property and decreases induced acute kidney injury in mice.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 35356-70-8
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.14
MDL Number MFCD00013394
InChI Key SMWNFFKPVLVOQQ-UHFFFAOYSA-N
Synonym methyl 2-acetamidoacrylate, 2-acetamidoacrylic acid methyl ester, n-acetyldehydroalanine methyl ester, 2-propenoic acid, 2-acetylamino-, methyl ester, methyl 2-acetylamino acrylate, 2-acetamidomethacrylate, acmc-209igf, methyl 2-acetaminoacrylate, methyl 2-acetylamino-acrylate, methyl 2-acetamido-2-propenate
PubChem CID 98644
IUPAC Name methyl 2-acetamidoprop-2-enoate
SMILES COC(=O)C(=C)NC(C)=O

Specifications

Melting Point 49°C to 51°C
Boiling Point 103°C to 104°C (8 mmHg)
Quantity 5 g
Beilstein 1812208
Formula Weight 143.14
Percent Purity 99%
Chemical Name or Material Methyl 2-acetamidoacrylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only