accessibility menu, dialog, popup

UserName

CAS RN 35303-76-5

NH2SNH2OO

CAS RN 35303-76-5

IUPAC Name: 4-(2-aminoethyl)benzene-1-sulfonamide
Synonyms: 4-(2-aminoethyl)benzenesulfonamide
Molecular Weight (g/mol): 200.26
Molecular Formula: C8H12N2O2S
InChi Key: FXNSVEQMUYPYJS-UHFFFAOYSA-N
SMILES: NCCC1=CC=C(C=C1)S(N)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results