accessibility menu, dialog, popup

UserName

O(E)(E)

CAS RN 35225-79-7

IUPAC Name: (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one
Synonyms: dibenzylideneacetone
Molecular Weight (g/mol): 234.3
Molecular Formula: C17H14O
InChi Key: WMKGGPCROCCUDY-PHEQNACWSA-N
SMILES: O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)