accessibility menu, dialog, popup

UserName

CAS RN 352-13-6

IUPAC Name: bromo(4-fluorophenyl)magnesium
Synonyms: bromo(4-fluorophenyl)magnesium
Molecular Weight (g/mol): 199.31
Molecular Formula: C6H4BrFMg
InChi Key: QYBFFRXNNFXREA-UHFFFAOYSA-M
SMILES: FC1=CC=C([Mg]Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results