accessibility menu, dialog, popup

UserName

CH3OOClCl

CAS RN 35112-28-8

IUPAC Name: methyl 2,4-dichlorobenzoate
Synonyms: methyl 2,4-dichlorobenzoate
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: VCRWILYAWSRHBN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results