accessibility menu, dialog, popup

UserName

CAS RN 3496-11-5

IUPAC Name: 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoate|Z-VAL-OSU
Molecular Weight (g/mol): 348.36
Molecular Formula: C17H20N2O6
InChi Key: MFAOBGXYLNLLJE-HNNXBMFYSA-N
SMILES: CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)