missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzoxy-L-valine Succinimidyl Ester 98.0+%, TCI America™

Catalog No. C23345G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C23345G Supplier TCI America Supplier No. C23345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3496-11-5
Molecular Formula C17H20N2O6
Molecular Weight (g/mol) 348.36
MDL Number MFCD00053547
InChI Key MFAOBGXYLNLLJE-HNNXBMFYSA-N
Synonym z-val-osu, n-cbz-l-valine succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino-3-methylbutanoate, l-valine,n-phenylmethoxy carbonyl-, 2,5-dioxo-1-pyrrolidinyl ester, zvalosu, 2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-3-methylbutanoate, cbz-val-osu, ksc496o1j, benzyl s-1-2,5-dioxo-1-pyrrolidinyl oxy carbonyl-2-methylpropyl carbamate, n-cbz-valinyl oxy-succinimide
PubChem CID 853556
IUPAC Name 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoate
SMILES CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 116°C
Color White
Quantity 5 g
Formula Weight 348.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-valine Succinimidyl Ester
TSCA No
Recommended Storage Freezer