missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, 98%

Catalog No. AAH6325314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6325314 25 g
AAH6325306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6325314 Supplier Thermo Scientific Chemicals Supplier No. H6325314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3496-11-5 | C17H27ClN8O5 | 458.90 g/mol

N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, is an amino acid which is widely used in pharmaceutical and food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-L-valine N-succinimidyl ester, is an amino acid which is widely used in pharmaceutical and food industry.

Solubility
Soluble in 1mmol in 2mL DCM.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 3496-11-5
MDL Number MFCD00053547

Specifications

Melting Point 114°C to 118°C
Color White
Molecular Formula C17H20N2O6
Beilstein 1555287
Synonym z-val-osu, n-cbz-l-valine succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino-3-methylbutanoate, l-valine,n-phenylmethoxy carbonyl-, 2,5-dioxo-1-pyrrolidinyl ester, zvalosu, 2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-3-methylbutanoate, cbz-val-osu, ksc496o1j, benzyl s-1-2,5-dioxo-1-pyrrolidinyl oxy carbonyl-2-methylpropyl carbamate, n-cbz-valinyl oxy-succinimide
Solubility Information Soluble in 1mmol in 2mL DCM.
InChI Key MFAOBGXYLNLLJE-HNNXBMFYSA-N
SMILES CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoate
Quantity 25 g
Molecular Weight (g/mol) 348.36
PubChem CID 853556
Formula Weight 348.35
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Benzyloxycarbonyl-L-valine N-succinimidyl ester
Show More Show Less
EINECSNumber 222-493-3
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only