accessibility menu, dialog, popup

UserName

CAS RN 349-59-7

IUPAC Name: 1-ethenyl-3,5-bis(trifluoromethyl)benzene
Synonyms: 1-ethenyl-3,5-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 240.15
Molecular Formula: C10H6F6
InChi Key: LFICVUCVPKKPFF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(C=C)=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results