CAS RN 346-06-5
IUPAC Name:
[2-(trifluoromethyl)phenyl]methanol
Synonyms:
[2-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol):
176.14
Molecular Formula:
C8H7F3O
InChi Key:
TWQNSHZTQSLJEE-UHFFFAOYSA-N
SMILES:
OCC1=CC=CC=C1C(F)(F)F
1
–
1
of
1
results
1
2-(Trifluoromethyl)benzyl Alcohol 98.0+%, TCI America™
CAS: 346-06-5 Molecular Formula: C8H7F3O Molecular Weight (g/mol): 176.14 MDL Number: MFCD00004621 InChI Key: TWQNSHZTQSLJEE-UHFFFAOYSA-N Synonym: 2-trifluoromethyl benzyl alcohol,2-trifluoromethyl phenyl methanol,o-trifluoromethylbenzyl alcohol,2-trifluoromethyl benzylalcohol,2-trifluoromethylbenzyl alcohol,o-trifluoromethyl benzyl alcohol,2-trifluoromethyl benzylic alcohol,benzenemethanol, 2-trifluoromethyl,2-trifluoromethyl phenyl methan-1-ol PubChem CID: 67666 IUPAC Name: [2-(trifluoromethyl)phenyl]methanol SMILES: OCC1=CC=CC=C1C(F)(F)F