accessibility menu, dialog, popup

UserName

H2OH2OH2OCaH3CCH3N(R)OH(R)OHOOFONHH3CCH3N(R)OH(R)OHOOFONH

CAS RN 344423-98-9

IUPAC Name: calcium bis((3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-(propan-2-yl)-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoate) trihydrate
Synonyms: sortis cardyl totalip atorvastatin calcium trihydrate torvast
Molecular Weight (g/mol): 1209.41
Molecular Formula: C66H74CaF2N4O13
InChi Key: SHZPNDRIDUBNMH-NIJVSVLQSA-L
SMILES: O.O.O.[Ca++].CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1.CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)