accessibility menu, dialog, popup

UserName

CAS RN 3440-28-6

HOONHO

CAS RN 3440-28-6

IUPAC Name: 3-(phenylformamido)propanoic acid
Synonyms: β-alanine, N-benzoyl-
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: CWXYHOHYCJXYFQ-UHFFFAOYSA-N
SMILES: OC(=O)CCNC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results