missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoyl-beta-alanine 98.0+%, TCI America™
$87.60 - $257.24
Chemical Identifiers
| CAS | 3440-28-6 |
|---|---|
| Molecular Formula | C10H11NO3 |
| Molecular Weight (g/mol) | 193.202 |
| MDL Number | MFCD00037293 |
| InChI Key | CWXYHOHYCJXYFQ-UHFFFAOYSA-N |
| Synonym | Betamipron |
| PubChem CID | 71651 |
| IUPAC Name | 3-benzamidopropanoic acid |
| SMILES | C1=CC=C(C=C1)C(=O)NCCC(=O)O |
Chemical Identifiers
| 3440-28-6 | |
| 193.202 | |
| CWXYHOHYCJXYFQ-UHFFFAOYSA-N | |
| 71651 | |
| C1=CC=C(C=C1)C(=O)NCCC(=O)O |
| C10H11NO3 | |
| MFCD00037293 | |
| Betamipron | |
| 3-benzamidopropanoic acid |
Specifications
| 3440-28-6 | |
| White | |
| MFCD00037293 | |
| Betamipron | |
| C1=CC=C(C=C1)C(=O)NCCC(=O)O | |
| 193.202 | |
| 193.20 | |
| Crystalline Powder |
| 132°C | |
| C10H11NO3 | |
| 5 g | |
| CWXYHOHYCJXYFQ-UHFFFAOYSA-N | |
| 3-benzamidopropanoic acid | |
| 71651 | |
| ≥98.0% (HPLC,T) | |
| Benzoyl-beta-alanine |
Safety and Handling
TSCA : No