missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluoroacetone trihydrate, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 18°C to 21°C |
| Density | 1.579 |
| Boiling Point | 105°C to 106°C |
| Refractive Index | 1.319 |
| Quantity | 100 g |
| UN Number | UN2552 |
| Beilstein | 607236 |
| Formula Weight | 220.07 (166.02 Anhydrous) |
| Percent Purity | 98% |
| Chemical Name or Material | Hexafluoroacetone trihydrate |
Safety and Handling
| Hazard Category | H301-H310+H330-H315-H318-H335-H361 |
| Hazard Statement | GHS H Statement H301-H310-H330-H318-H361-H315-H335 Toxic if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes serious eye damage. Suspected of damaging fertility or the unborn child. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P302+P350-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: HEXAFLUOROACETONE HYDRATE, LIQUID |
| EINECSNumber | 211-676-3 |
| RTECSNumber | UC2700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only