missing translation for 'onlineSavingsMsg'
Learn More

Hexafluoroacetone Hydrate 97.0+%, TCI America™

Catalog No. H04255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04255G 5 g
H042525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04255G Supplier TCI America Supplier No. H04255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34202-69-2
Molecular Formula C3H6F6O4
Molecular Weight (g/mol) 220.067
MDL Number MFCD00149061
InChI Key SNZAEUWCEHDROX-UHFFFAOYSA-N
Synonym hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate
PubChem CID 36719
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
SMILES C(=O)(C(F)(F)F)C(F)(F)F.O.O.O

Specifications

Color Yellow
Quantity 5 g
UN Number 2552
Formula Weight 166.02
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Hexafluoroacetone Hydrate
EINECSNumber (2)-0581
RTECSNumber UC2700000
TSCA No