accessibility menu, dialog, popup

UserName

H3CONH2OOH

CAS RN 3403-47-2

IUPAC Name: 5-amino-2-methoxybenzoic acid
Synonyms: 5-amino-2-methoxybenzoic acid
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: LWWPSEIFAKNPKQ-UHFFFAOYSA-N
SMILES: COC1=C(C=C(N)C=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results