accessibility menu, dialog, popup

UserName

CH3OOOOCH3

CAS RN 3399-22-2

IUPAC Name: 1,4-dimethyl cyclohexane-1,4-dicarboxylate
Synonyms: 1,4-dimethyl cyclohexane-1,4-dicarboxylate
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: LNGAGQAGYITKCW-UHFFFAOYSA-N
SMILES: COC(=O)C1CCC(CC1)C(=O)OC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)