missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl trans-1,4-Cyclohexanedicarboxylate 96.0+%, TCI America™

Catalog No. D37235G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37235G 5 g
D37231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37235G Supplier TCI America Supplier No. D37235G
Only null left

Chemical Identifiers

CAS 3399-22-2
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
MDL Number MFCD00063917,MFCD00001460,MFCD00216473
InChI Key LNGAGQAGYITKCW-UHFFFAOYSA-N
Synonym dimethyl 1,4-cyclohexanedicarboxylate, dimethyl trans-1,4-cyclohexanedicarboxylate, 1r,4r-dimethyl cyclohexane-1,4-dicarboxylate, cis-dimethyl cyclohexane-1,4-dicarboxylate, dimethyl trans-cyclohexane-1,4-dicarboxylate, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, dimethyl hexahydroterephthalate, trans-dimethyl cyclohexane-1,4-dicarboxylate, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, trans, unii-ys87mds0zv
PubChem CID 7198
IUPAC Name 1,4-dimethyl cyclohexane-1,4-dicarboxylate
SMILES COC(=O)C1CCC(CC1)C(=O)OC

Specifications

Melting Point 69°C
Color White
Boiling Point 140°C
Quantity 5 g
Formula Weight 200.23
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl trans-1,4-Cyclohexanedicarboxylate
EINECSNumber (3)-2435&(3)-2449
TSCA Yes