accessibility menu, dialog, popup

UserName

H3COOCH3NOONOO

CAS RN 3395-03-7

IUPAC Name: 1,2-dimethoxy-4,5-dinitrobenzene
Synonyms: 1,2-dimethoxy-4,5-dinitrobenzene
Molecular Weight (g/mol): 228.16
Molecular Formula: C8H8N2O6
InChi Key: WFDHPWTYKOAFBJ-UHFFFAOYSA-N
SMILES: COC1=CC(=C(C=C1OC)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results