CAS RN 33893-89-9
IUPAC Name:
5-(pyridin-2-yl)-1H-1,2,3,4-tetrazol-1-ide
Synonyms:
5-(pyridin-2-yl)-1H-1,2,3,4-tetrazol-1-ide
Molecular Weight (g/mol):
146.13
Molecular Formula:
C6H4N5
InChi Key:
ZKHJXYLAESJIEI-UHFFFAOYSA-N
SMILES:
[N-]1N=NN=C1C1=CC=CC=N1
1
–
1
of
1
results
1
Promotions Available
5-(2-Pyridyl)-1H-tetrazole, 98%
5-(2-Pyridyl)-1H-tetrazole, 98%
CAS: 33893-89-9 Molecular Formula: C6H4N5 Molecular Weight (g/mol): 146.13 MDL Number: MFCD00068114 InChI Key: ZKHJXYLAESJIEI-UHFFFAOYSA-N Synonym: 5-2-pyridyl-1h-tetrazole,2-1h-tetrazol-5-yl pyridine,2-2h-tetrazol-5-yl pyridine,2-2h-tetrazol-5-yl-pyridine,pyridine, 2-1h-tetrazol-5-yl,5-2-pyridinyl tetrazole,2-1h-1,2,3,4-tetrazol-5-yl pyridine,5-2-pyridyl tetrazole,5-pyridin-2-yl tetrazole,chembl80094 PubChem CID: 320267 SMILES: [N-]1N=NN=C1C1=CC=CC=N1