missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(2-Pyridyl)-1H-tetrazole, 98%
Description
It is used in the synthesis and crystal structure of the [Ag(2-allyl-5-phenyl-2Htetrazole)ClO4] π,complex .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis and crystal structure of the [Ag(2-allyl-5-phenyl-2Htetrazole)ClO4] π,σ-complex .
Solubility
Soluble in water at 25°C 11 g/L.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used in the synthesis and crystal structure of the [Ag(2-allyl-5-phenyl-2Htetrazole)ClO4] π,σ-complex .
Solubility
Soluble in water at 25°C 11 g/L.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Quantity | 5 g |
| Solubility Information | Soluble in water at 25°C 11g/L. |
| IUPAC Name | 2-(2H-tetrazol-5-yl)pyridine |
| Formula Weight | 147.14 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | 5-(2-Pyridyl)-1H-tetrazole |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only